ENA321765321 Enamine

2-methylpentane-3-sulfonamide

  • MDL number MFCD16672077
  • Cas Number : 1247155-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76670

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76670
Availability details for
Quantity
  • Availability
  • Add to Cart