ENA321765324 Enamine

2-(cyclopentyloxy)propanoic acid

  • MDL number MFCD12145313
  • Cas Number : 854673-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76672

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76672
Availability details for
Quantity
  • Availability
  • Add to Cart