ENA321765332 Enamine

N-(1-carbamothioylcyclopentyl)benzamide

  • MDL number MFCD13339675
  • Cas Number : 1274493-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-76677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76677
Availability details for
Quantity
  • Availability
  • Add to Cart