ENA321765563 Enamine

N-(2-chloroethyl)cyclohexanamine hydrochloride

  • MDL number MFCD18839086
  • Cas Number : 50597-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77822
Availability details for
Quantity
  • Availability
  • Add to Cart