ENA321765572 Enamine

2-amino-3,3-dimethylbutan-1-ol hydrochloride

  • MDL number MFCD18839093
  • Cas Number : 1306604-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77893

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77893
Availability details for
Quantity
  • Availability
  • Add to Cart