ENA321765576 Enamine

2-chloro-4-fluoro-N'-hydroxybenzene-1-carboximidamide hydrochloride

  • Cas Number : 1311319-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77923
Availability details for
Quantity
  • Availability
  • Add to Cart