ENA321765582 Enamine

3-bromo-1-(1-methyl-1H-pyrazol-4-yl)piperidin-2-one

  • MDL number MFCD18839097
  • Cas Number : 1306606-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77959
Availability details for
Quantity
  • Availability
  • Add to Cart