ENA321765594 Enamine

6-bromo-2-methylpyrazolo[1,5-a]pyrimidine-3-sulfonyl chloride

  • MDL number MFCD18839107
  • Cas Number : 1306606-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77993

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77993
Availability details for
Quantity
  • Availability
  • Add to Cart