ENA321765596 Enamine

6-bromopyrazolo[1,5-a]pyrimidine-3-carbaldehyde

  • MDL number MFCD18839109
  • Cas Number : 1263060-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77996

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-77996
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart