ENA321765607 Enamine

5-(4-bromophenyl)-1,2,4-oxadiazole

  • MDL number MFCD18839120
  • Cas Number : 89892-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78018
Availability details for
Quantity
  • Availability
  • Add to Cart