ENA321765608 Enamine

5-(3-bromophenyl)-1,2,4-oxadiazole

  • MDL number MFCD18839121
  • Cas Number : 1306606-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78019
Availability details for
Quantity
  • Availability
  • Add to Cart