ENA321765625 Enamine

1-amino-3-(2-aminoethyl)urea dihydrochloride

  • MDL number MFCD18917298
  • Cas Number : 1311318-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78077
Availability details for
Quantity
  • Availability
  • Add to Cart