ENA321765629 Enamine

2,2,2-trifluoroethyl N-(3-chloro-1-benzothiophen-2-yl)carbamate

  • MDL number MFCD18917302
  • Cas Number : 1311313-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-78094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78094
Availability details for
Quantity
  • Availability
  • Add to Cart