ENA321765633 Enamine

2-methoxy-6-(piperazin-1-yl)pyrazine

  • MDL number MFCD18917303
  • Cas Number : 89007-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78110
Availability details for
Quantity
  • Availability
  • Add to Cart