ENA321765637 Enamine

1-(3-bromopropyl)-1H-imidazole hydrobromide

  • MDL number MFCD18917307
  • Cas Number : 109914-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78134

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-78134
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart