ENA321765645 Enamine

1-[2-(4-methoxyphenyl)-1-methylpiperidin-3-yl]methanamine

  • MDL number MFCD18917315
  • Cas Number : 1311315-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78207
Availability details for
Quantity
  • Availability
  • Add to Cart