ENA321765647 Enamine

6-(2-oxo-1,3-oxazolidin-3-yl)pyridine-3-carbonitrile

  • MDL number MFCD18917317
  • Cas Number : 1311318-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78274

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78274
Availability details for
Quantity
  • Availability
  • Add to Cart