ENA321765650 Enamine

({3,5-dimethyl-4-[(4-methylphenyl)methoxy]phenyl}methyl)(methyl)amine hydrochloride

  • MDL number MFCD18917321
  • Cas Number : 1311318-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78303
Availability details for
Quantity
  • Availability
  • Add to Cart