ENA321765651 Enamine

({3,5-dimethyl-4-[(4-methylphenyl)methoxy]phenyl}methyl)(ethyl)amine hydrochloride

  • MDL number MFCD18917322
  • Cas Number : 1311315-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-78304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78304
Availability details for
Quantity
  • Availability
  • Add to Cart