ENA321765652 Enamine

2-amino-2-(2,6-difluorophenyl)ethan-1-ol hydrochloride

  • MDL number MFCD18917323
  • Cas Number : 1311317-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232...234
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78306

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78306
Availability details for
Quantity
  • Availability
  • Add to Cart