ENA321765658 Enamine

1-[3-(3-chlorophenyl)-1,2,4-oxadiazol-5-yl]-6-azaspiro[2.5]octane hydrochloride

  • MDL number MFCD18917330
  • Cas Number : 1311318-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78353
Availability details for
Quantity
  • Availability
  • Add to Cart