ENA321765659 Enamine

3-[4-(2-chloroacetyl)phenyl]propanamide

  • MDL number MFCD18917331
  • Cas Number : 341553-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-78355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78355
Availability details for
Quantity
  • Availability
  • Add to Cart