ENA321765660 Enamine

2-methyl-5-(4-methyl-1,3-thiazol-2-yl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD18917332
  • Cas Number : 1311315-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78365
Availability details for
Quantity
  • Availability
  • Add to Cart