ENA321765662 Enamine

1-({3-[(dimethylamino)methyl]phenyl}sulfanyl)-N,N-dimethylformamide

  • MDL number MFCD18917334
  • Cas Number : 1311318-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78384
Availability details for
Quantity
  • Availability
  • Add to Cart