ENA321765665 Enamine

2-(4-methoxyphenyl)-1-methyl-6-oxopiperidine-3-carboxamide

  • MDL number MFCD18917337
  • Cas Number : 1311316-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220...222
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78419

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78419
Availability details for
Quantity
  • Availability
  • Add to Cart