ENA321765667 Enamine

({3,5-dimethyl-4-[(3-methylphenyl)methoxy]phenyl}methyl)(methyl)amine hydrochloride

  • MDL number MFCD18917339
  • Cas Number : 1311316-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-78461

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78461
Availability details for
Quantity
  • Availability
  • Add to Cart