ENA321765668 Enamine

({3,5-dimethyl-4-[(3-methylphenyl)methoxy]phenyl}methyl)(ethyl)amine hydrochloride

  • MDL number MFCD18917340
  • Cas Number : 1311317-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-78462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78462
Availability details for
Quantity
  • Availability
  • Add to Cart