ENA321765673 Enamine

4-methyl-3-(pyridin-4-yloxy)benzoic acid hydrochloride

  • MDL number MFCD18917345
  • Cas Number : 1311315-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78513
Availability details for
Quantity
  • Availability
  • Add to Cart