ENA321765680 Enamine

1-[(1-phenyl-1H-pyrazol-4-yl)methyl]piperazine dihydrochloride

  • MDL number MFCD18917352
  • Cas Number : 1311316-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78590

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78590
Availability details for
Quantity
  • Availability
  • Add to Cart