ENA321765683 Enamine

4-ethyl-6-methyl-1H,6H-[1,2]diazolo[3,4-c]pyrazol-3-amine

  • MDL number MFCD18887014
  • Cas Number : 1311318-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78600
Availability details for
Quantity
  • Availability
  • Add to Cart