ENA321765689 Enamine

[3-(1H-imidazol-1-yl)phenyl]methanamine

  • MDL number MFCD18917359
  • Cas Number : 1311315-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78642
Availability details for
Quantity
  • Availability
  • Add to Cart