ENA321765693 Enamine

methyl 2-bromo-4-fluoro-5-sulfamoylbenzoate

  • MDL number MFCD18897523
  • Cas Number : 1311318-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78658
Availability details for
Quantity
  • Availability
  • Add to Cart