ENA321765702 Enamine

N-(piperidin-4-yl)pyridazin-3-amine dihydrochloride

  • MDL number MFCD18917373
  • Cas Number : 1311315-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78758
Availability details for
Quantity
  • Availability
  • Add to Cart