ENA321765706 Enamine

1-methyl-2-(1-methyl-1H-pyrazol-4-yl)-6-oxopiperidine-3-carboxylic acid

  • MDL number MFCD18917376
  • Cas Number : 1311316-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225...227
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78773

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78773
Availability details for
Quantity
  • Availability
  • Add to Cart