ENA321765709 Enamine

4-[(2-bromophenyl)sulfanyl]butan-1-amine hydrochloride

  • MDL number MFCD18917379
  • Cas Number : 1311316-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-78804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78804
Availability details for
Quantity
  • Availability
  • Add to Cart