ENA321765710 Enamine

2-(1-phenylethoxy)aniline hydrochloride

  • MDL number MFCD18917380
  • Cas Number : 1311317-13-1
  • Molecular Weight : 249.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-78816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78816
Availability details for
Quantity
  • Availability
  • Add to Cart