ENA321765713 Enamine

[4-(4-chlorophenyl)-1H-imidazol-2-yl]methanamine dihydrochloride

  • MDL number MFCD18917383
  • Cas Number : 1311317-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259...261
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78840

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78840
Availability details for
Quantity
  • Availability
  • Add to Cart