ENA321765714 Enamine

3-bromo-1-(3-fluorophenyl)piperidin-2-one

  • MDL number MFCD18917384
  • Cas Number : 1311316-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78846
Availability details for
Quantity
  • Availability
  • Add to Cart