ENA321765718 Enamine

1-methyl-6-oxo-2-[4-(trifluoromethyl)phenyl]piperidine-3-carboxylic acid

  • MDL number MFCD07377693
  • Cas Number : 1311316-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78928
Availability details for
Quantity
  • Availability
  • Add to Cart