ENA321765719 Enamine

[2-(4-methoxyphenyl)-1-methylpyrrolidin-3-yl]methanol

  • MDL number MFCD18917388
  • Cas Number : 75810-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78934
Availability details for
Quantity
  • Availability
  • Add to Cart