ENA321765720 Enamine

N,N-dimethyl-1-{[1-(4-methylphenyl)-1H-pyrazol-4-yl]sulfanyl}formamide

  • MDL number MFCD18917389
  • Cas Number : 1311314-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78944
Availability details for
Quantity
  • Availability
  • Add to Cart