ENA321765723 Enamine

N-(3-methyl-2-oxobutyl)-4-nitrobenzamide

  • MDL number MFCD18917393
  • Cas Number : 1311317-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-78972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78972
Availability details for
Quantity
  • Availability
  • Add to Cart