ENA321765736 Enamine

4-[2-(methylamino)ethoxy]phenol hydrochloride

  • MDL number MFCD18917406
  • Cas Number : 857198-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-79127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79127
Availability details for
Quantity
  • Availability
  • Add to Cart