ENA321765738 Enamine

4-(2-bromoethyl)-1-methyl-1H-pyrazole hydrobromide

  • MDL number MFCD18917408
  • Cas Number : 1311317-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79139

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-79139
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart