ENA321765742 Enamine

2-[(2,2-dimethylpropyl)amino]-1-phenylethan-1-ol hydrochloride

  • MDL number MFCD18917412
  • Cas Number : 1311318-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79182
Availability details for
Quantity
  • Availability
  • Add to Cart