ENA321765750 Enamine

5-methyl-3-sulfanyl-[1,2,4]triazolo[3,4-b][1,3]thiazole-6-carboxylic acid

  • MDL number MFCD18917421
  • Cas Number : 1311316-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-79389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79389
Availability details for
Quantity
  • Availability
  • Add to Cart