ENA321765774 Enamine

2-(2-oxopropyl)nicotinic acid

  • MDL number MFCD18839140
  • Cas Number : 35969-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78111

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78111
Availability details for
Quantity
  • Availability
  • Add to Cart