ENA321765776 Enamine

ethyl N-(2-amino-1-cyclopropylethyl)carbamate

  • MDL number MFCD16707023
  • Cas Number : 1247926-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78115

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78115
Availability details for
Quantity
  • Availability
  • Add to Cart