ENA321765780 Enamine

2-[(3-hydroxyphenyl)formamido]acetamide

  • MDL number MFCD11130775
  • Cas Number : 1019353-72-0
  • Molecular Weight : 194.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-78127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78127
Availability details for
Quantity
  • Availability
  • Add to Cart