ENA321765782 Enamine

2,2,6-trimethylheptan-3-amine

  • MDL number MFCD11655433
  • Cas Number : 1099683-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77899

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77899
Availability details for
Quantity
  • Availability
  • Add to Cart